missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Samplicity™ G2 Filtration System

Catalog No. SAMP2SYSG
Change view
Click to view available options
Color:
Bold Blue
Glossy Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
SAMP2SYSG Glossy Green
SAMP2SYSB Bold Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SAMP2SYSG Supplier MilliporeSigma™ Supplier No. SAMP2SYSG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vacuum filtration system using Millex™ filters for sample preparation for HPLC.

Overview of Samplicity™ G2 System
  • Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
  • Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
  • Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration

Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC

Order Info

Does not include Millex filters.

Specifications

Color Glossy Green
Content And Storage Base, Lid, Vial Tray, Waste Tray, Tubing
Material Polypropylene with Polyester and Silicone Components
For Use With (Application) Analytical sample prep; High Performance Liquid Chromatography (HPLC)
Housing Material PVDF
Includes Base, Lid, Vial Tray, Waste Tray, Tubing
Quantity 1 Kit
Dimensions (L x W x H) 44.5 x 10.2 x 9.1 cm
Diameter (Metric) Filter 33 mm
For Use With (Equipment) Millex Filter (Non-sterile)