missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Acid Washed Grinding Beads
Acid Washed Grinding Beads are treated to remove fine particles and contaminants.
$867.98 - $867.98
Specifications
| Product Type | Grinding Beads |
|---|---|
| For Use With (Application) | Grinding |
| For Use With (Equipment) | 2010 Geno/Grinder |
| Packaging Quantity | 200 g |
| Particle Size | 800 to 1000 μm |
Description
- Choose from silica or zirconia grinding beads to help remove fine particles and contaminant.
- Using untreated beads which contain impurities can affect homogenization and biological processes.
- Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples.
- Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power.
- General-purpose grinding beads are suitable for most basic cell lysis applications.
- Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes.
- Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.
Specifications
| Grinding Beads | |
| 2010 Geno/Grinder | |
| 800 to 1000 μm |
| Grinding | |
| 200 g |