Learn More
MilliporeSigma™ Supelco™ Sample Loop for Model 7725 Injector, Stainless Steel
Catalog No. 111004849
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
50 μL
100 μL
200 μL
1 mL
2 mL
5 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
111004849 5 μL
111004945 10 μL
111004999 20 μL
111004946 50 μL
111004930 100 μL
111004881 200 μL
111004718 1 mL
111004510 2 mL
111004178 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 111004849 Supplier MilliporeSigma™ Supelco™ Supplier No. 57623
Sample loop for model 7725 injector (volume 100 μL) is best suited for advanced fluid-handling solutions for broad range of analytical instrumentation and in vitro diagnostic systems. It is useful in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Specifications
Material Stainless Steel
For Use With (Equipment) Model 7725
Volume (Metric) 5 μL