missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
GREENER_CHOICE

Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.

$422.50 - $458.69

Specifications

Product Type Specimen Container
Closure Included Yes
Material Virgin Polypropylene
Shape Cylindrical
Capacity (Metric) 120 mL
View More Specs

Products 3
Catalog Number Mfr. No. Sterility Closure Color Label No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Closure Color Label No. per Pack Price Quantity & Availability  
1371156
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
01 0001
Sterile Orange Tabbed 75/Pk., Pre-assembled
Case of 300 Each for $458.69
Only null left
 
1371157
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
01 0100
Non-sterile Yellow Non-tabbed 75/Pk., Pre-assembled
Case of 300 Each for $429.13
Only null left
 
22610146
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
N01 0195
Non-sterile Orange No Label 300/Pk., Bagged Separately
Case of 300 Each for $422.50
Only null left
 
Description

Description

Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:

  • 120mL (4 oz.) 53mm
  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Stackable containers are easy to store, saving lab space
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:
Case of 300

Specifications

Specifications

Specimen Container
Virgin Polypropylene
120 mL
Bio-Tite Lid
Yes
Cylindrical
53 mm
53 mm
Videos
SDS
Documents

Documents