Learn More
Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers

Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Supplier: Thermo Scientific™ 01 0100
Description
Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:
- 120mL (4 oz.) 53mm
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- Dual-threaded caps create improved sealing
- Sterilized by gamma irradiation, which eliminates potential contamination
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Stackable containers are easy to store, saving lab space
- Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Ordering Information:
Case of 300
Chemical Identifiers
| Specimen Container | |
| Virgin Polypropylene | |
| 120 mL | |
| Bio-Tite Lid |
| Yes | |
| Cylindrical | |
| 53 mm | |
| 53 mm |
Specifications
| Specimen Container | |
| Virgin Polypropylene | |
| Non-sterile | |
| Yellow | |
| Bio-Tite Lid | |
| 53 mm | |
| 300/Cs. |
| Yes | |
| Cylindrical | |
| 120 mL | |
| 53 mm | |
| Cap and container pre-assembled | |
| Non-tabbed | |
| 75/Pk., Pre-assembled |