missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Salmeterol xinafoate

Catalog No. 471210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
471210 10 mg
471250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 471210 Supplier Tocris Bioscience™ Supplier No. 4712/10*

Long-acting β2 agonist; bronchodilator

Long-acting β2 adrenergic receptor agonist. Inhibits the release of histamine, leukotrienes and prostaglandin D2, and blocks GM-CSF production from human cultured mast cells. Exhibits efficacious bronchodilatory activity in patients with asthma or chronic obstructive pulmonary disease (COPD).

Chemical Identifiers

CAS 94749-08-3
Molecular Formula C36H45NO7

Specifications

Quantity 10 mg
Formula Weight 603.75
Purity >99%
Molecular Weight (g/mol) 603.76
Chemical Name or Material Salmeterol xinafoate
Recommended Storage Desiccate at Room Temperature