missing translation for 'onlineSavingsMsg'
Learn More

Salicylhydroxamic acid, 99%

Catalog No. p-7081507
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1152809 10 g
AAA1152814 25 g
AAA1152818 50 g
AAA1152822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1152809 Supplier Thermo Scientific Chemicals Supplier No. A1152809
Only null left

CAS: 89-73-6 | C7H7NO3 | 153.137 g/mol

It is widely used for a variety of roles in biology and medicine as a chelating therapy. It inhibits bacterial or fungi growth by interfering with iron uptake. It is also active as a inhibitor of enzyme involved in tumour growths. These compounds are used as extractants for the separation of valuable metals or minerals including copper, lead, gold, other rare earth metals and kaolin. Hydroxamic acids are also used to produce corresponding isocyanates by dehydration followed by rearrangement reaction. It is also used as a common ligand utilized in the synthesis of metallacrowns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used for a variety of roles in biology and medicine as a chelating therapy. It inhibits bacterial or fungi growth by interfering with iron uptake. It is also active as a inhibitor of enzyme involved in tumour growths. These compounds are used as extractants for the separation of valuable metals or minerals including copper, lead, gold, other rare earth metals and kaolin. Hydroxamic acids are also used to produce corresponding isocyanates by dehydration followed by rearrangement reaction. It is also used as a common ligand utilized in the synthesis of metallacrowns.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong bases. Stable under normal conditions.

Chemical Identifiers

CAS 89-73-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00002110
InChI Key HBROZNQEVUILML-UHFFFAOYSA-N
Synonym salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide
PubChem CID 66644
ChEBI CHEBI:45615
IUPAC Name N,2-dihydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NO)O

Specifications

Melting Point ∼170°C (decomposition)
Quantity 10 g
Beilstein 1210520
Merck Index 14,8331
Solubility Information It is soluble in water.
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material Salicylhydroxamic acid
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 201-934-3
RTECSNumber VO6870000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only