missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide 98.0+%, TCI America™

Catalog No. S000625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000625G 25 g
S0006500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000625G Supplier TCI America Supplier No. S000625G

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007978
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
PubChem CID 5147
ChEBI CHEBI:32114
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)C1=CC=CC=C1O

Specifications

Melting Point 140°C
Color White
Boiling Point 182°C
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Salicylamide
EINECSNumber (3)-1571
RTECSNumber VN6475000
TSCA Yes