accessibility menu, dialog, popup

UserName

CAS RN 65-45-2

IUPAC Name: 2-hydroxybenzamide
Synonyms: salicylamide O-hydroxybenzamide OHB salicilamida 2-carboxamidophenol salicylamidum
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: SKZKKFZAGNVIMN-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)