Learn More
Salicylamide, 98%
CAS: 65-45-2 | C7H7NO2 | 137.14 g/mol
Supplier: Thermo Scientific Chemicals A107970B
Description
Salicylamide is used in combination with both aspirin and caffeine in the over-the-counter pain remedies. It is used as an analgesic and as an antipyretic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSalicylamide is used in combination with both aspirin and caffeine in the over-the-counter pain remedies. It is used as an analgesic and as an antipyretic.
Solubility
Soluble in water (2.06 mg/ml at 25°C), chloroform, dimethyl sulfoxide, alcohol, and methanol (100 mg/ml).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| Salicylamide | |
| 138°C to 142°C | |
| C7H7NO2 | |
| 1000 g | |
| 14,8328 | |
| Soluble in water (2.06mg/ml at 25°C),chloroform,dimethyl sulfoxide,alcohol,and methanol (100mg/ml). | |
| NC(=O)C1=CC=CC=C1O | |
| 137.14 | |
| CHEBI:32114 | |
| 98% |
| 65-45-2 | |
| 181°C (14 mmHg) | |
| MFCD00007978 | |
| 742439 | |
| salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin | |
| SKZKKFZAGNVIMN-UHFFFAOYSA-N | |
| 2-hydroxybenzamide | |
| 5147 | |
| 137.14 |
Chemical Identifiers
| 65-45-2 | |
| 137.14 | |
| SKZKKFZAGNVIMN-UHFFFAOYSA-N | |
| 5147 | |
| 2-hydroxybenzamide |
| C7H7NO2 | |
| MFCD00007978 | |
| salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin | |
| CHEBI:32114 | |
| NC(=O)C1=CC=CC=C1O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 200-609-3
RTECSNumber : VN6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only