missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salbutamol Hemisulfate 98.0+%, TCI America™
$63.52 - $444.72
Chemical Identifiers
| CAS | 51022-70-9 |
|---|---|
| Molecular Formula | C13H23NO7S |
| Molecular Weight (g/mol) | 337.387 |
| MDL Number | MFCD00055200 |
| InChI Key | OVICLFZZVQVVFT-UHFFFAOYSA-N |
| Synonym | salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate |
| PubChem CID | 9884233 |
| IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
| SMILES | CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O |
Chemical Identifiers
| 51022-70-9 | |
| 337.387 | |
| OVICLFZZVQVVFT-UHFFFAOYSA-N | |
| 9884233 | |
| CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O |
| C13H23NO7S | |
| MFCD00055200 | |
| salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate | |
| 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
Specifications
| 51022-70-9 | |
| White-Yellow | |
| MFCD00055200 | |
| salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate | |
| CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O | |
| 337.387 | |
| 288.35 | |
| Crystalline Powder |
| 180°C | |
| C13H23NO7S | |
| 1 g | |
| OVICLFZZVQVVFT-UHFFFAOYSA-N | |
| 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid | |
| 9884233 | |
| ≥98.0% (HPLC,T) | |
| Salbutamol Hemisulfate |
Safety and Handling
RTECSNumber : CZ6430100
TSCA : No