missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol hemisulfate, 98+%

Catalog No. AAA1854403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1854403 1 g
AAA1854406 5 g
AAA1854414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1854403 Supplier Thermo Scientific Chemicals Supplier No. A1854403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51022-70-9 | C13H23NO7S | 337.387 g/mol

Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

Solubility
Soluble in water and 1M sodium hydroxide (50 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 51022-70-9
Molecular Formula C13H23NO7S
Molecular Weight (g/mol) 337.387
MDL Number MFCD00055200
InChI Key OVICLFZZVQVVFT-UHFFFAOYSA-N
Synonym salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
PubChem CID 9884233
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
SMILES CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O

Specifications

Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in water and 1M sodium hydroxide (50mg/ml).
Formula Weight 576.7
Percent Purity 99%
Chemical Name or Material Salbutamol sulfate
Hazard Category H317-H351-H361fd
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 256-916-8
RTECSNumber CZ6430100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only