missing translation for 'onlineSavingsMsg'
Learn More

StatLab Sakura Cyto-Tek Base Holder

Numéro de catalogue. 21033443
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Material
21033443 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 21033443 Fournisseur StatLab Code fournisseur STLM963B
Il en reste null

Disposable component used in a cytocentrifuge to hold the fluid chamber and filter paper during the process of preparing cell samples for slides

  • Plastic
  • CS200
  • It is a disposable item made of a material like polypropylene.
  • Its main purpose is to facilitate the preparation of consistent, high-quality cell samples from liquid specimens.
  • It is used in conjunction with a fluid chamber and filter paper in Sakura Cyto-Tek centrifuges for creating monolayer slides from non-gynecological samples.

Spécifications

Color White
For Use With (Equipment) Sakura Cyto-Tek
Material Plastic
For Use With (Application) Cytocentrifuge
Product Line Base Holder
Length (Metric) 107.95 cm
Width (Metric) 101.6 cm