missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ PTFE Griffin Beakers
Chemware™ PTFE Griffin Beakers are noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid.
$291.64 - $1,136.25
Specifications
| Material | PTFE |
|---|---|
| For Use With (Application) | Ideal for Digestion |
| Includes | Spout |
| Temperature (English) Max. Operating | 550°F |
| Temperature (Metric) Max. Operating | 288°C |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
025861F
|
Saint Gobain Life Sciences
D1069036 |
50 mL |
Each for $291.64
|
|
|||||
025861K
|
Saint Gobain Life Sciences
D1069040 |
600 mL |
Each for $892.68
|
|
|||||
025861M
|
Saint Gobain Life Sciences
D1069041 |
1000 mL |
Each for $1,136.25
|
|
|||||
Description
- Nonwettable, unbreakable and heat resistant
- Easy to clean
- Broad spout for easy pouring
- Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
- Specially molded bottom allows rapid heat transfer
- Unlike most PTFE beakers, these will boil water when covered
Applications:
Ideal for digestion
Specifications
| PTFE | |
| Spout | |
| 288°C | |
| Griffin |
| Ideal for Digestion | |
| 550°F | |
| Beaker |