missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ PTFE Griffin Beakers

Catalog No. 025861F
Encompass
Change view
Click to view available options
Capacity (Metric):
50 mL
600 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
025861F 50 mL
025861K 600 mL
025861M 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 025861F Supplier Saint Gobain Life Sciences Supplier No. D1069036

Chemware™ PTFE Griffin Beakers are noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid.

  • Nonwettable, unbreakable and heat resistant
  • Easy to clean
  • Broad spout for easy pouring
  • Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
  • Specially molded bottom allows rapid heat transfer
  • Unlike most PTFE beakers, these will boil water when covered

Applications:

Ideal for digestion

Specifications

Capacity (English) 1.69 oz.
Capacity (Metric) 50 mL
Material PTFE
For Use With (Application) Ideal for Digestion
Height (English) 2.05 in.
Height (Metric) 5.23 cm
Includes Spout
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C
Type Beaker
Style Griffin
Show More Show Less