missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ PTFE Griffin Beakers
Chemware™ PTFE Griffin Beakers are noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid.
Supplier: Saint Gobain Life Sciences D1069041
Description
- Nonwettable, unbreakable and heat resistant
- Easy to clean
- Broad spout for easy pouring
- Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
- Specially molded bottom allows rapid heat transfer
- Unlike most PTFE beakers, these will boil water when covered
Applications:
Ideal for digestion
Chemical Identifiers
| PTFE | |
| Spout | |
| 288°C | |
| Griffin |
| Ideal for Digestion | |
| 550°F | |
| Beaker |
Specifications
| 33.81 oz. | |
| PTFE | |
| 5.75 in. | |
| Spout | |
| 288°C | |
| Griffin |
| 1000 mL | |
| Ideal for Digestion | |
| 14.62 cm | |
| 550°F | |
| Beaker |