missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SAG dihydrochloride
Description
SAG dihydrochloride is a water-soluble dihydrochloride salt of SAG (Cat. No. 4366). SAG is a potent Smoothened (Smo) receptor agonist (Kd = 59 nM); antagonizes Cyclopamine (Cat. No. 1623) action at the Smo receptor. SAG potently activates the Hedgehog signaling pathway in Shh-light 2 cells (EC50 ∼ 3 nM) and induces pathway activation independently of Ptch proteins. SAG is a putative inhibitor of a cellular component required for Hedgehog signaling and also enhances neuronal differentiation of iPSCs into dopaminergic neurons.
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]benzo[b]thiophene-2-carboxamide dihydrochloride |
| CAS | 2702366-44-5 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Product Line | Tocris |
| Molecular Formula | C28H28ClN3OS0.2 HCl |
| Formula Weight | Observed MW: 562.98 |
| Purity | 98% |