Learn More
Fisherbrand™ Safety Coated Clear Boston Round Glass Bottles without Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage with plastisol coating for safety are easier to grip and harder to break.
 View more versions of this product
Catalog No. FB02911921
Only null left
Description
Description
  • Coated in layer of plastisol
  • Shatter-resistant coating reduces chance of breakage
  • If broken, safety coating will contain contents long enough for disposal
  • Non-slip coating also allows greater ease of stability, wet or dry
  • Not autoclavable
Specifications
Chemical Identifiers
Clear
No
Safety-coated Boston Round Bottle
Glass (Type 3)
Narrow
Specifications
480 mL
Clear
16 mm
60
No
168 mm
28-400 mm
16 oz.
25 mm
Glass (Type 3)
28-400
Narrow
Safety-coated Boston Round Bottle
75 mm
Product Suggestions
Product Suggestions