missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saccharin Sodium, Dihydrate, Crystalline, USP, 99-101%, Spectrum™ Chemical
SO195, 6155-57-3, C7H4NNaO3S·2H2O
Supplier: Spectrum Chemical Mfg Cor SO19512KG
Description
Spectrum™ Chemical Saccharin Sodium, Dihydrate, Crystalline, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 6155-57-3 | |
| Poly Pail | |
| 12 kg | |
| O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 | |
| 241.19 | |
| USP |
| 1 | |
| C7H8NNaO5S | |
| NAUHOSQQKJTUAX-UHFFFAOYSA-M | |
| sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate | |
| 99 to 101% |
Chemical Identifiers
| 6155-57-3 | |
| 241.19 | |
| sodium 1,3-dioxo-3H-1λâ¶,2-benzothiazol-1-olate dihydrate |
| C7H8NNaO5S | |
| NAUHOSQQKJTUAX-UHFFFAOYSA-M | |
| O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |