missing translation for 'onlineSavingsMsg'
Learn More

Saccharin Sodium, Dihydrate, Crystalline, USP, 99-101%, Spectrum™ Chemical

Catalog No. p-7158142
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606118 100 g Poly Bottle
18606114 500 g Poly Bottle
18606115 2.5 kg Poly Bottle
18606116 12 kg Poly Pail
18606117 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606118 Supplier Spectrum Chemical Mfg Cor Supplier No. SO195100GM
Only null left

SO195, 6155-57-3, C7H4NNaO3S·2H2O

Spectrum™ Chemical Saccharin Sodium, Dihydrate, Crystalline, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6155-57-3
Molecular Formula C7H8NNaO5S
Molecular Weight (g/mol) 241.19
InChI Key NAUHOSQQKJTUAX-UHFFFAOYSA-M
IUPAC Name sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate
SMILES O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 99 to 101%
Grade USP