missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-XylBINAP 97.0+%, TCI America™

Catalog No. p-7137976
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0071200MG 200 mg
X00711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X0071200MG Supplier TCI America Supplier No. X0071200MG

Chemical Identifiers

CAS 135139-00-3
Molecular Formula C52H48P2
Molecular Weight (g/mol) 734.904
MDL Number MFCD01630821
InChI Key MXGXXBYVDMVJAO-UHFFFAOYSA-N
Synonym s-dm-binap, dm-binap, r-dm-binap, s---2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, s-3,5-xylyl-binap, r-+-2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, 3,5-xylyl-binap, s-xylyl-binap, r-2,2'-bis bis 3,5-dimethylphenyl phosphino-1,1'-binaphthalene, r-+-xylbinap
PubChem CID 4189905
IUPAC Name [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C

Specifications

Melting Point 255°C
Color White-Yellow
Quantity 200 mg
Formula Weight 734.90
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-XylBINAP