accessibility menu, dialog, popup

UserName

CH3PPCH3H3CH3CCH3CH3H3CH3C(?)

CAS RN 135139-00-3

IUPAC Name: {2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane
Synonyms: {2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol): 734.9
Molecular Formula: C52H48P2
InChi Key: MXGXXBYVDMVJAO-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results