missing translation for 'onlineSavingsMsg'
Learn More

S-Trityl-L-cysteinamide, 98%

Catalog No. p-7055126
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6362803 1 g
AAH6362806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6362803 Supplier Thermo Scientific Chemicals Supplier No. H6362803
Only null left

CAS: 166737-85-5 | C22H22N2OS | 362.491 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 166737-85-5
Molecular Formula C22H22N2OS
Molecular Weight (g/mol) 362.491
MDL Number MFCD22126061
InChI Key OHWBGKONMFYEKL-FQEVSTJZSA-N
Synonym h-cys trt-nh2, r-2-amino-3-tritylthio propanamide, s-trityl-l-cysteinamide, s-trityl-l-cysteine amide, s-trityl-l-cysteinamide hydrochloride, 2r-2-amino-3-triphenylmethyl sulfanyl propanamide, propanamide,2-amino-3-triphenylmethyl thio-, 2r, r-2-amino-3-tritylthio propanamide hydrochloride, ambotzhaa1560, h-cys trt-nh2.hcl
PubChem CID 44432703
IUPAC Name (2R)-2-amino-3-tritylsulfanylpropanamide
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)N)N

Specifications

Color White
Quantity 1 g
Formula Weight 362.49
Percent Purity 98%
Physical Form Powder
Chemical Name or Material S-Trityl-L-cysteinamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only