missing translation for 'onlineSavingsMsg'
Learn More

(S)-tert-Leucinol, 95%

CAS: 112245-13-3 | C6H16NO | 118.20 g/mol

$523.32

Chemical Identifiers

CAS 112245-13-3
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00192250
InChI Key JBULSURVMXPBNA-RXMQYKEDSA-O
Synonym l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
PubChem CID 2734079
SMILES CC(C)(C)[C@H]([NH3+])CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC377530050
View Documents Promotion Details
N/A 5 g Glass bottle
Each for $523.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112245-13-3
118.20
JBULSURVMXPBNA-RXMQYKEDSA-O
2734079
C6H16NO
MFCD00192250
l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
CC(C)(C)[C@H]([NH3+])CO

Specifications

Authentic
112245-13-3
100.0
90g/mL
Crystalline Low Melting Mass or Liquid
C6H16NO
MFCD00192250
JBULSURVMXPBNA-RXMQYKEDSA-O
(2S)-1-hydroxy-3,3-dimethylbutan-2-aminium
2734079
≥94%
White or Colorless
94.0
+34° to +40° (25°C, 589nm) (c=1.5, EtOH)
+ 37.50 (20.00°C c=2,EtOH)
95%
(CH3)3CCH(NH2)CH2OH
l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
CC(C)(C)[C@H]([NH3+])CO
118.20
117.19
(S)-tert-Leucinol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only