accessibility menu, dialog, popup

UserName

H3CCH3CH3(S)NH2OH

CAS RN 112245-13-3

IUPAC Name: (2S)-1-hydroxy-3,3-dimethylbutan-2-aminium
Synonyms: (2S)-1-hydroxy-3,3-dimethylbutan-2-aminium
Molecular Weight (g/mol): 118.2
Molecular Formula: C6H16NO
InChi Key: JBULSURVMXPBNA-RXMQYKEDSA-O
SMILES: CC(C)(C)[C@H]([NH3+])CO
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)