missing translation for 'onlineSavingsMsg'
Learn More

S,S'-Dimethyl Dithiocarbonate 98.0+%, TCI America™

Catalog No. D21145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21145G Supplier TCI America Supplier No. D21145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 868-84-8
Molecular Formula C3H6OS2
Molecular Weight (g/mol) 122.20
MDL Number MFCD00144163
InChI Key IUXMJLLWUTWQFX-UHFFFAOYSA-N
Synonym Dithiocarbonic Acid S,S′C-Dimethyl Ester
PubChem CID 313474
IUPAC Name bis(methylsulfanyl)methanone
SMILES CSC(=O)SC

Specifications

Color Yellow
Boiling Point 169°C
Quantity 5 g
Formula Weight 122.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S,S′-Dimethyl Dithiocarbonate