accessibility menu, dialog, popup

UserName

CAS RN 868-84-8

IUPAC Name: bis(methylsulfanyl)methanone
Synonyms: bis(methylsulfanyl)methanone
Molecular Weight (g/mol): 122.2
Molecular Formula: C3H6OS2
InChi Key: IUXMJLLWUTWQFX-UHFFFAOYSA-N
SMILES: CSC(=O)SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results