missing translation for 'onlineSavingsMsg'
Learn More

S,S'-Diethyl Dithiocarbonate 98.0+%, TCI America™

Catalog No. D207810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D207810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D207810G Supplier TCI America Supplier No. D207810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 623-80-3
Molecular Formula C5H10OS2
Molecular Weight (g/mol) 150.25
MDL Number MFCD00144162
InChI Key MBUCFVKJCBBWRI-UHFFFAOYSA-N
Synonym Dithiocarbonic Acid S,S′C-Diethyl Ester
PubChem CID 12196
IUPAC Name bis(ethylsulfanyl)methanone
SMILES CCSC(=O)SCC

Specifications

Color Yellow
Boiling Point 168°C
Quantity 10 g
Formula Weight 150.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S,S′-Diethyl Dithiocarbonate