accessibility menu, dialog, popup

UserName

H3CSOSCH3

CAS RN 623-80-3

IUPAC Name: bis(ethylsulfanyl)methanone
Synonyms: S,S-diethyl carbonodithioate
Molecular Weight (g/mol): 150.25
Molecular Formula: C5H10OS2
InChi Key: MBUCFVKJCBBWRI-UHFFFAOYSA-N
SMILES: CCSC(=O)SCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results