missing translation for 'onlineSavingsMsg'
Learn More

S-p-Tolyl p-Toluenesulfonothioate 98.0+%, TCI America™

Catalog No. T31731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31731G 1 g
T31735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31731G Supplier TCI America Supplier No. T31731G

Chemical Identifiers

CAS 2943-42-2
Molecular Formula C14H14O2S2
Molecular Weight (g/mol) 278.38
MDL Number MFCD00092126
InChI Key HSAROCRPZOYKGS-UHFFFAOYSA-N
Synonym p-Toluenesulfonothioic Acid S-p-Tolyl Ester
PubChem CID 257647
IUPAC Name 1-methyl-4-[(4-methylbenzenesulfonyl)sulfanyl]benzene
SMILES CC1=CC=C(SS(=O)(=O)C2=CC=C(C)C=C2)C=C1

Specifications

Melting Point 74°C
Color White
Quantity 1 g
Formula Weight 278.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material S-p-Tolyl p-Toluenesulfonothioate