missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-O-Desmethylraclopride 98.0+%, TCI America™
$105.89 - $371.56
Chemical Identifiers
| CAS | 119670-11-0 |
|---|---|
| Molecular Formula | C14H18Cl2N2O3 |
| Molecular Weight (g/mol) | 333.209 |
| MDL Number | MFCD02179385 |
| InChI Key | DPQOMEPZWBXAMA-QMMMGPOBSA-N |
| Synonym | (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide |
| PubChem CID | 10947574 |
| IUPAC Name | 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dihydroxybenzamide |
| SMILES | CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2O)Cl)Cl)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
D4134200MG
|
TCI America
D4134200MG |
200 mg |
Each for $105.89
|
|
|||||
|
D41341G
|
TCI America
D41341G |
1 g |
Each for $371.56
|
|
|||||
Chemical Identifiers
| 119670-11-0 | |
| 333.209 | |
| DPQOMEPZWBXAMA-QMMMGPOBSA-N | |
| 10947574 | |
| CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2O)Cl)Cl)O |
| C14H18Cl2N2O3 | |
| MFCD02179385 | |
| (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide | |
| 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dihydroxybenzamide |
Specifications
| 119670-11-0 | |
| White-Yellow | |
| MFCD02179385 | |
| (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide | |
| CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2O)Cl)Cl)O | |
| 333.209 | |
| 333.21 | |
| Crystalline Powder |
| 223°C | |
| C14H18Cl2N2O3 | |
| 200 mg | |
| DPQOMEPZWBXAMA-QMMMGPOBSA-N | |
| 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dihydroxybenzamide | |
| 10947574 | |
| ≥98.0% (HPLC,T) | |
| (S)-O-Desmethylraclopride |
Safety and Handling
TSCA : No