missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-O-Desmethylraclopride 98.0+%, TCI America™
Supplier: TCI America D41341G
Specifications
| (S)-O-Desmethylraclopride | |
| 223°C | |
| C14H18Cl2N2O3 | |
| 1 g | |
| DPQOMEPZWBXAMA-QMMMGPOBSA-N | |
| 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dihydroxybenzamide | |
| 10947574 | |
| ≥98.0% (HPLC,T) |
| 119670-11-0 | |
| White-Yellow | |
| MFCD02179385 | |
| (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide | |
| CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2O)Cl)Cl)O | |
| 333.209 | |
| 333.21 | |
| Crystalline Powder |
Chemical Identifiers
| 119670-11-0 | |
| 333.209 | |
| DPQOMEPZWBXAMA-QMMMGPOBSA-N | |
| 10947574 | |
| CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2O)Cl)Cl)O |
| C14H18Cl2N2O3 | |
| MFCD02179385 | |
| (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide | |
| 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dihydroxybenzamide |
Safety and Handling
TSCA : No