missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%

Catalog No. AAH5197503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197503 1 g
AAH5197506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197503 Supplier Thermo Scientific Chemicals Supplier No. H5197503

CAS: 136030-33-6 | C25H21NO4 | 399.446 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136030-33-6
Molecular Formula C25H21NO4
Molecular Weight (g/mol) 399.446
MDL Number MFCD00144368
InChI Key LIRBCUNCXDZOOU-QHCPKHFHSA-N
Synonym fmoc-tic-oh, fmoc-l-tic, fmoc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-2-9h-fluoren-9-yl methoxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-+-2-9-fluorenylmethoxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, n-fmoc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-9h-fluoren-9-ylmethoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, 3s-2-9h-fluoren-9-ylmethoxy carbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, fmoc-tic, pubchem6028
PubChem CID 978341
IUPAC Name (3S)-2-(9H-fluoren-9-ylmethoxycarbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMILES C1C(N(CC2=CC=CC=C21)C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O

Specifications

Quantity 1 g
Beilstein 4827574
Optical Rotation +26.0° (c=1 in Methanol)
Formula Weight 399.45
Percent Purity 95%
Chemical Name or Material (S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only