missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-Benzyl-1-phenylethylamine 98.0+%, TCI America™

Catalog No. p-7119096
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15295ML 5 mL
B152925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15295ML Supplier TCI America Supplier No. B15295ML

Chemical Identifiers

CAS 17480-69-2
Molecular Formula C15H24N
Molecular Weight (g/mol) 218.36
MDL Number MFCD00066325
InChI Key UHABCGJJMSQRRP-ZDUSSCGKSA-O
Synonym s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, benzyl 1s-1-phenylethyl amine, s-n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, pubchem5973, n-benzyl-d-amphetamine
PubChem CID 1268085
IUPAC Name (cyclohexylmethyl)[(1S)-1-phenylethyl]azanium
SMILES C[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 211.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-N-Benzyl-1-phenylethylamine