accessibility menu, dialog, popup

UserName

H3C(S)NH2

CAS RN 17480-69-2

IUPAC Name: (cyclohexylmethyl)[(1S)-1-phenylethyl]azanium
Synonyms: (cyclohexylmethyl)[(1S)-1-phenylethyl]azanium
Molecular Weight (g/mol): 218.36
Molecular Formula: C15H24N
InChi Key: UHABCGJJMSQRRP-ZDUSSCGKSA-O
SMILES: C[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)