missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%

Catalog No. AAL1929906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1929906 5 g
AAL1929914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1929906 Supplier Thermo Scientific Chemicals Supplier No. L1929906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17480-69-2 | C15H24N | 218.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17480-69-2
Molecular Formula C15H24N
Molecular Weight (g/mol) 218.36
MDL Number MFCD00066325
InChI Key UHABCGJJMSQRRP-ZDUSSCGKSA-O
Synonym s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, benzyl 1s-1-phenylethyl amine, s-n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, pubchem5973, n-benzyl-d-amphetamine
PubChem CID 1268085
IUPAC Name (1S)-N-benzyl-1-phenylethanamine
SMILES C[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1

Specifications

Density 1.01
Boiling Point 171°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.564
Quantity 5 g
Beilstein 3650264
Sensitivity Air Sensitive
Optical Rotation −39° (Neat)
Formula Weight 211.31
Percent Purity 99%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-N-Benzyl-1-phenylethylamine
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 605-736-1
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only