missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl methanethiolsulfonate, 97%

CAS: 2949-92-0 | C2H6O2S2 | 126.19 g/mol

$77.04 - $77.04

Chemical Identifiers

CAS 2949-92-0
Molecular Formula C2H6O2S2
Molecular Weight (g/mol) 126.19
MDL Number MFCD00007565
InChI Key XYONNSVDNIRXKZ-UHFFFAOYSA-N
Synonym s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane
PubChem CID 18064
ChEBI CHEBI:74357
IUPAC Name methylsulfonylsulfanylmethane
SMILES CSS(=O)(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC197390010
SDS View Documents
Thermo Scientific Chemicals
197390010
1 g Glass bottle
Each for $77.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2949-92-0
126.19
XYONNSVDNIRXKZ-UHFFFAOYSA-N
18064
methylsulfonylsulfanylmethane
C2H6O2S2
MFCD00007565
s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane
CHEBI:74357
CSS(=O)(=O)C

Specifications

2949-92-0
69.0°C to 71.0°C (0.4 mmHg)
96% min. (GC)
C2H6O2S2
CH3SO2SCH3
1 g
1.227
XYONNSVDNIRXKZ-UHFFFAOYSA-N
methylsulfonylsulfanylmethane
18064
126.19
S-Methyl methanethiolsulfonate
1.2270g/mL
87°C
Glass bottle
1.5118
MFCD00007565
05,454; 07,243; 12,325; 01,169; 10,107; 16,99
s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane
CSS(=O)(=O)C
126.19
CHEBI:74357
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Warning

EINECSNumber : 220-970-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only