missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl Trityl Ether 98.0+%, TCI America™

$123.03 - $361.62

Chemical Identifiers

CAS 129940-50-7
Molecular Formula C22H20O2
Molecular Weight (g/mol) 316.4
MDL Number MFCD00273373
InChI Key XFSXUCMYFWZRAF-NRFANRHFSA-N
Synonym s---trityl glycidyl ether, s-2-trityloxy methyl oxirane, s-trityl glycidyl ether, s---glycidyl trityl ether, s-glycidyl trityl ether, 2s-2-trityloxymethyl oxirane, s-tritylglycidylether, s-2-triphenylmethoxymethyl oxirane, s---glycidyl triphenylmethyl ether, s-glycidyl triphenylmethyl ether
PubChem CID 7168113
IUPAC Name (2S)-2-(trityloxymethyl)oxirane
SMILES C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G02855G
View Documents
TCI America
G02855G
5 g
Each for $123.03
Only null left
 
G028525G
View Documents
TCI America
G028525G
25 g
Each for $361.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

129940-50-7
316.4
XFSXUCMYFWZRAF-NRFANRHFSA-N
7168113
C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
C22H20O2
MFCD00273373
s---trityl glycidyl ether, s-2-trityloxy methyl oxirane, s-trityl glycidyl ether, s---glycidyl trityl ether, s-glycidyl trityl ether, 2s-2-trityloxymethyl oxirane, s-tritylglycidylether, s-2-triphenylmethoxymethyl oxirane, s---glycidyl triphenylmethyl ether, s-glycidyl triphenylmethyl ether
(2S)-2-(trityloxymethyl)oxirane

Specifications

129940-50-7
White-Yellow
MFCD00273373
s---trityl glycidyl ether, s-2-trityloxy methyl oxirane, s-trityl glycidyl ether, s---glycidyl trityl ether, s-glycidyl trityl ether, 2s-2-trityloxymethyl oxirane, s-tritylglycidylether, s-2-triphenylmethoxymethyl oxirane, s---glycidyl triphenylmethyl ether, s-glycidyl triphenylmethyl ether
C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
316.4
316.40
Crystalline Powder
101°C
C22H20O2
5 g
XFSXUCMYFWZRAF-NRFANRHFSA-N
(2S)-2-(trityloxymethyl)oxirane
7168113
≥98.0% (GC)
(S)-Glycidyl Trityl Ether
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'