accessibility menu, dialog, popup

UserName

O(S)O

CAS RN 129940-50-7

IUPAC Name: (2S)-2-[(triphenylmethoxy)methyl]oxirane
Synonyms: (2S)-2-[(triphenylmethoxy)methyl]oxirane
Molecular Weight (g/mol): 316.4
Molecular Formula: C22H20O2
InChi Key: XFSXUCMYFWZRAF-NRFANRHFSA-N
SMILES: C(OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[C@@H]1CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results