missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl Trityl Ether 98.0+%, TCI America™

Catalog No. G02855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02855G 5 g
G028525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02855G Supplier TCI America Supplier No. G02855G
Only null left

Chemical Identifiers

CAS 129940-50-7
Molecular Formula C22H20O2
Molecular Weight (g/mol) 316.4
MDL Number MFCD00273373
InChI Key XFSXUCMYFWZRAF-NRFANRHFSA-N
Synonym s---trityl glycidyl ether, s-2-trityloxy methyl oxirane, s-trityl glycidyl ether, s---glycidyl trityl ether, s-glycidyl trityl ether, 2s-2-trityloxymethyl oxirane, s-tritylglycidylether, s-2-triphenylmethoxymethyl oxirane, s---glycidyl triphenylmethyl ether, s-glycidyl triphenylmethyl ether
PubChem CID 7168113
IUPAC Name (2S)-2-(trityloxymethyl)oxirane
SMILES C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 316.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-Glycidyl Trityl Ether