missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl 4-Nitrobenzenesulfonate 98.0+%, TCI America™

Catalog No. g04115g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G04115G Supplier TCI America Supplier No. G04115G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 118712-60-0
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.232
MDL Number MFCD08460170
InChI Key CXYDYDCHYJXOEY-QMMMGPOBSA-N
Synonym 4-Nitrobenzenesulfonic Acid (S)-Glycidyl Ester
PubChem CID 10868995
IUPAC Name [(2S)-oxiran-2-yl]methyl 4-nitrobenzenesulfonate
SMILES C1C(O1)COS(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 259.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-Glycidyl 4-Nitrobenzenesulfonate