missing translation for 'onlineSavingsMsg'
Learn More

S-Ethylisothiourea Hydrobromide 98.0+%, TCI America™

Catalog No. p-7128882
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E018225G 25 g
E0182100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E018225G Supplier TCI America Supplier No. E018225G
Only null left

Chemical Identifiers

CAS 1071-37-0
Molecular Formula C3H9BrN2S
Molecular Weight (g/mol) 185.083
MDL Number MFCD00012585
InChI Key SWXXKWPYNMZFTE-UHFFFAOYSA-N
Synonym s-ethylisothiourea hydrobromide, 2-ethyl-2-thiopseudourea hydrobromide, ethiron, ethiron bromide, s-ethylthiuronium bromide, s-ethylisothiouronium hydrobromide, bromide s-ethylisothiuronium, s-ethylisothiuronium bromide, usaf el-18, isothuron hydrobromide
PubChem CID 200213
IUPAC Name ethyl carbamimidothioate;hydrobromide
SMILES CCSC(=N)N.Br

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 25 g
Formula Weight 185.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-Ethylisothiourea Hydrobromide
EINECSNumber (2)-2866
RTECSNumber UM6825000
TSCA No