accessibility menu, dialog, popup

UserName

CAS RN 1071-37-0

IUPAC Name: (ethylsulfanyl)methanimidamide hydrobromidyl
Synonyms: ethylisothiourea hydrobromidyl
Molecular Weight (g/mol): 184.08
Molecular Formula: C3H8BrN2S
InChi Key: KZGYLFSEGYUERO-UHFFFAOYSA-N
SMILES: [Br].CCSC(N)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results