missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Curve™ Beta Storage Containers
Ideal for storing reagents that emit beta isotopes
$420.20 - $520.11
Specifications
| Color | Clear |
|---|---|
| Material | Acrylic |
| For Use With (Application) | For storing reagents that emit beta isotopes |
| Includes | Rubber pads, flame treated edges |
| Product Type | Beta storage container |
Description
- 3/8 in. thick (0.95cm) clear acrylic provides a protective beta barrier
- Full-length hinge for durability and strength
- Rubber pads on bottom to prevent sliding and scratching
- Choice of two sizes
Specifications
| Clear | |
| For storing reagents that emit beta isotopes | |
| Beta storage container |
| Acrylic | |
| Rubber pads, flame treated edges | |
| 0.37 in. |