missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Citalopram Oxalate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 146°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 414.43 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (S)-Citalopram Oxalate |
Safety and Handling
| RTECSNumber | NP6333500 |
| TSCA | No |
| Recommended Storage | Refrigerator |