missing translation for 'onlineSavingsMsg'
Learn More

(S)-Citalopram Oxalate 98.0+%, TCI America™

Catalog No. E0958200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0958200MG 200 mg
E09581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0958200MG Supplier TCI America Supplier No. E0958200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 219861-08-2
Molecular Formula C22H23FN2O5
Molecular Weight (g/mol) 414.433
MDL Number MFCD06407826
InChI Key KTGRHKOEFSJQNS-BDQAORGHSA-N
Synonym escitalopram oxalate, lexapro, cipralex, s-citalopram oxalate, citalopram oxalate, unii-5u85dbw7lo, 5u85dbw7lo, s-+-citalopram oxalate, s-1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile oxalate
PubChem CID 146571
IUPAC Name (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.C(=O)(C(=O)O)O

Specifications

Melting Point 146°C
Color White
Quantity 200 mg
Formula Weight 414.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-Citalopram Oxalate
RTECSNumber NP6333500
TSCA No
Recommended Storage Refrigerator