missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™

Catalog No. M08321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08321G 1 g
M08325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08321G Supplier TCI America Supplier No. M08321G

Chemical Identifiers

CAS 17257-71-5
Molecular Formula C10H8F3O3
Molecular Weight (g/mol) 233.17
MDL Number MFCD00064200
InChI Key JJYKJUXBWFATTE-VIFPVBQESA-M
Synonym s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
PubChem CID 6992788
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoate
SMILES CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Color White
Quantity 1 g
Formula Weight 234.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic Acid [Optical Resolving]