missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane 97.0+%, TCI America™
Supplier: TCI America C17965G
Specifications
| (S)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane | |
| Colorless | |
| C6H11ClO2 | |
| 5 g | |
| s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s, s-3-chloro-1,2-propanediol acetonide, pubchem14076, bnpotxlwpzoesz-rxmqykedsa | |
| CC1(OCC(O1)CCl)C | |
| 150.602 | |
| 150.60 | |
| Liquid |
| 60456-22-6 | |
| 63°C | |
| MFCD00273365 | |
| 1992 | |
| BNPOTXLWPZOESZ-RXMQYKEDSA-N | |
| (4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane | |
| 6930483 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 60456-22-6 | |
| 150.602 | |
| BNPOTXLWPZOESZ-RXMQYKEDSA-N | |
| 6930483 | |
| CC1(OCC(O1)CCl)C |
| C6H11ClO2 | |
| MFCD00273365 | |
| s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane, s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane, 1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s, s-3-chloro-1,2-propanediol acetonide, pubchem14076, bnpotxlwpzoesz-rxmqykedsa | |
| (4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane |
Safety and Handling
RTECSNumber : JH6950000
TSCA : No