accessibility menu, dialog, popup

UserName

CH3H3CO(R)ClO

CAS RN 60456-22-6

IUPAC Name: (4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Synonyms: (4R)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: BNPOTXLWPZOESZ-YFKPBYRVSA-N
SMILES: CC1(C)OC[C@H](CCl)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results