missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyloxy-1,2-butanediol 95.0+%, TCI America™

Catalog No. p-7119833
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2900200MG 200 mg
B29001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2900200MG Supplier TCI America Supplier No. B2900200MG

Chemical Identifiers

CAS 69985-32-6
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.25
MDL Number MFCD10000950
InChI Key TVRPDIKPMQUOSL-UHFFFAOYNA-N
PubChem CID 9794180
IUPAC Name 4-(benzyloxy)butane-1,2-diol
SMILES OCC(O)CCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 170°C
Quantity 200 mg
Formula Weight 196.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-4-Benzyloxy-1,2-butanediol